Assay Details
Gene Symbol: Mat2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044282.1 | NP_001037747.1 | 6 - 7 | 911 | 96 | |
XM_006246126.2 | XP_006246188.1 | 6 - 7 | 1061 | 96 | ||
GenBank mRNA | BC085899.1 | - | 6 - 7 | 911 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
