Assay Details
Gene Symbol: Gnptg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100493.1 | NP_001093963.1 | 4 - 5 | 248 | 66 | |
XM_006245935.2 | XP_006245997.1 | 4 - 5 | 265 | 66 | ||
GenBank mRNA | BC098716.1 | - | 4 - 5 | 230 | 66 | |
BC158818.1 | - | 4 - 5 | 248 | 66 | ||
FQ213320.1 | - | 4 - 5 | 255 | 66 | ||
FQ213586.1 | - | 4 - 5 | 213 | 66 | ||
FQ213702.1 | - | 4 - 5 | 247 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
