Assay Details
Gene Symbol: Gnptg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100493.1 | NP_001093963.1 | 7 - 8 | 544 | 56 | |
XM_006245935.2 | XP_006245997.1 | 7 - 8 | 561 | 56 | ||
GenBank mRNA | BC098716.1 | - | 7 - 8 | 526 | 56 | |
BC158818.1 | - | 7 - 8 | 544 | 56 | ||
FQ213320.1 | - | 7 - 8 | 551 | 56 | ||
FQ213586.1 | - | 7 - 8 | 509 | 56 | ||
FQ213702.1 | - | 7 - 8 | 543 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
