Assay Details
Gene Symbol: Gnptg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100493.1 | NP_001093963.1 | 9 - 10 | 759 | 82 | |
XM_006245935.2 | XP_006245997.1 | 9 - 10 | 776 | 82 | ||
GenBank mRNA | BC098716.1 | - | 9 - 10 | 741 | 82 | |
BC158818.1 | - | 9 - 10 | 759 | 82 | ||
FQ213320.1 | - | 9 - 10 | 766 | 82 | ||
FQ213586.1 | - | 9 - 10 | 724 | 82 | ||
FQ213702.1 | - | 9 - 10 | 759 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
