Assay Details
Gene Symbol: Rnf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770273.1 | XP_008768495.1 | 1 - 2 | 240 | 61 | |
GenBank mRNA | AY050655.1 | - | 1 - 2 | 132 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AY050655.1, XM_008770273.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
