Assay Details
Gene Symbol: Rnf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770272.1 | XP_008768494.1 | 1 - 2 | 111 | 70 | |
GenBank mRNA | AF022081.1 | - | 1 - 2 | 42 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF022081.1, XM_008770272.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
