Assay Details
Gene Symbol: Ctbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019201.3 | NP_062074.2 | 2 - 3 | 388 | 143 | |
XM_006251229.2 | XP_006251291.1 | 2 - 3 | 414 | 143 | ||
GenBank mRNA | AF067795.2 | - | 2 - 3 | 393 | 143 | |
BC078778.1 | - | 2 - 3 | 399 | 143 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF067795.2, BC078778.1, NM_019201.3, XM_006251229.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
