Assay Details
Gene Symbol: Ctbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019201.3 | NP_062074.2 | 4 - 5 | 686 | 129 | |
XM_006251225.2 | XP_006251287.1 | 3 - 4 | 521 | 129 | ||
XM_006251226.2 | XP_006251288.1 | 3 - 4 | 521 | 129 | ||
XM_006251227.2 | XP_006251289.1 | 3 - 4 | 520 | 129 | ||
XM_006251228.2 | XP_006251290.1 | 3 - 4 | 521 | 129 | ||
XM_006251229.2 | XP_006251291.1 | 4 - 5 | 712 | 129 | ||
GenBank mRNA | AF067795.2 | - | 4 - 5 | 691 | 129 | |
BC078778.1 | - | 4 - 5 | 697 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
