Assay Details
Gene Symbol: Rbpj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106631.1 | NP_001100101.1 | 9 - 10 | 904 | 91 | |
XM_008770190.1 | XP_008768412.1 | 8 - 9 | 1575 | 91 | ||
XM_008770191.1 | XP_008768413.1 | 8 - 9 | 816 | 91 | ||
XM_008770192.1 | XP_008768414.1 | 8 - 9 | 2994 | 91 | ||
XM_008770193.1 | XP_008768415.1 | 9 - 10 | 987 | 91 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
