Assay Details
Gene Symbol: Plk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107669.1 | NP_001101139.1 | 11 - 12 | 2354 | 116 | |
XM_006232299.2 | XP_006232361.1 | 12 - 13 | 2532 | 116 | ||
XM_006232300.1 | XP_006232362.1 | 11 - 12 | 2237 | 116 | ||
GenBank mRNA | BC166450.1 | - | 12 - 13 | 2472 | 8371068 | 116 |
FQ234784.1 | - | 8 - 9 | 1404 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
