Assay Details
Gene Symbol: Adad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006977.2 | NP_001006978.2 | 1 - 2 | 130 | 73 | |
XM_006232278.2 | XP_006232340.1 | 1 - 2 | 314 | 73 | ||
GenBank mRNA | BC105906.1 | - | 1 - 2 | 196 | 73 | |
CV126304.1 | - | 1 - 2 | 161 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
