Assay Details
Gene Symbol: Adad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006977.2 | NP_001006978.2 | 4 - 5 | 666 | 130 | |
XM_006232278.2 | XP_006232340.1 | 4 - 5 | 850 | 130 | ||
XM_006232279.2 | XP_006232341.1 | 5 - 6 | 702 | 130 | ||
GenBank mRNA | BC105906.1 | - | 4 - 5 | 732 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
