Assay Details
Gene Symbol: Adad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006977.2 | NP_001006978.2 | 6 - 7 | 857 | 142 | |
XM_006232278.2 | XP_006232340.1 | 6 - 7 | 1041 | 142 | ||
XM_006232279.2 | XP_006232341.1 | 7 - 8 | 893 | 142 | ||
GenBank mRNA | BC105906.1 | - | 6 - 7 | 923 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
