Assay Details
Gene Symbol: Il2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053836.1 | NP_446288.1 | 1 - 2 | 157 | 111 | |
GenBank mRNA | M22899.1 | - | 1 - 2 | 157 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
