Assay Details
Gene Symbol: Anxa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013132.1 | NP_037264.1 | 3 - 4 | 158 | 66 | |
XM_006232268.2 | XP_006232330.1 | 3 - 4 | 293 | 66 | ||
GenBank mRNA | AF051895.1 | - | 3 - 4 | 213 | 66 | |
BC081855.1 | - | 3 - 4 | 211 | 66 | ||
FQ213209.1 | - | 4 - 5 | 288 | 66 | ||
FQ214176.1 | - | 3 - 4 | 188 | 66 | ||
FQ229127.1 | - | 3 - 4 | 146 | 66 | ||
M21730.1 | - | 3 - 4 | 158 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
