Assay Details
Gene Symbol: Pms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105908.1 | NP_001099378.1 | 11 - 12 | 2141 | 109 | |
XM_006248862.2 | XP_006248924.1 | 12 - 13 | 2234 | 109 | ||
GenBank mRNA | BC160858.1 | - | 12 - 13 | 2173 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
