Assay Details
Gene Symbol: Pms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105908.1 | NP_001099378.1 | 12 - 13 | 2246 | 142 | |
XM_006248862.2 | XP_006248924.1 | 13 - 14 | 2339 | 142 | ||
GenBank mRNA | BC160858.1 | - | 13 - 14 | 2278 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
