Assay Details
Gene Symbol: Pms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105908.1 | NP_001099378.1 | 1 - 2 | 114 | 129 | |
XM_006248862.2 | XP_006248924.1 | 1 - 2 | 109 | 129 | ||
GenBank mRNA | BC160858.1 | - | 1 - 2 | 48 | 129 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC160858.1, NM_001105908.1, XM_006248862.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
