Assay Details
Gene Symbol: Pms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105908.1 | NP_001099378.1 | 8 - 9 | 977 | 116 | |
XM_006248862.2 | XP_006248924.1 | 9 - 10 | 1070 | 116 | ||
GenBank mRNA | BC160858.1 | - | 9 - 10 | 1009 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
