Assay Details
Gene Symbol: Xiap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022231.2 | NP_071567.1 | 6 - 7 | 1427 | 96 | |
XM_006257506.2 | XP_006257568.1 | 5 - 6 | 1253 | 96 | ||
XM_006257508.2 | XP_006257570.1 | 5 - 6 | 1367 | 96 | ||
XM_006257509.2 | XP_006257571.1 | 5 - 6 | 1268 | 96 | ||
XM_008773529.1 | XP_008771751.1 | 6 - 7 | 1394 | 96 | ||
GenBank mRNA | AB033366.1 | - | 6 - 7 | 1426 | 96 | |
AF183429.1 | - | 4 - 5 | 1097 | 96 | ||
AF304334.1 | - | 2022 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
