Assay Details
Gene Symbol: Stk26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191736.1 | NP_001178665.1 | 9 - 10 | 1233 | 69 | |
XM_006257573.2 | XP_006257635.1 | 11 - 12 | 1720 | 69 | ||
XM_008773567.1 | XP_008771789.1 | 10 - 11 | 2432 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
