Assay Details
Gene Symbol: Ube2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013933.1 | NP_001013955.1 | 6 - 7 | 495 | 83 | |
XM_006257446.2 | XP_006257508.1 | 5 - 6 | 508 | 83 | ||
GenBank mRNA | BC079353.1 | - | 6 - 7 | 495 | 7108236 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
