Assay Details
Gene Symbol: Ube2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013933.1 | NP_001013955.1 | 5 - 6 | 412 | 222 | |
XM_006257446.2 | XP_006257508.1 | 4 - 5 | 425 | 192 | ||
GenBank mRNA | BC079353.1 | - | 5 - 6 | 412 | 7108236 | 222 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
