Assay Details
Gene Symbol: Mbnl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106949.1 | NP_001100419.1 | 1 - 2 | 252 | 105 | |
XM_008773556.1 | XP_008771778.1 | 1 - 2 | 1731 | 105 | ||
XM_008773557.1 | XP_008771779.1 | 1 - 2 | 854 | 105 | ||
XM_008773558.1 | XP_008771780.1 | 1 - 2 | 851 | 105 | ||
XM_008773559.1 | XP_008771781.1 | 1 - 2 | 854 | 105 | ||
XM_008773560.1 | XP_008771782.1 | 1 - 2 | 857 | 105 | ||
XM_008773561.1 | XP_008771783.1 | 1 - 2 | 854 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
