Assay Details
Gene Symbol: Mbnl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106949.1 | NP_001100419.1 | 6 - 7 | 1084 | 79 | |
XM_008773558.1 | XP_008771780.1 | 7 - 8 | 1755 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P153671 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P153674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161447 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81262 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162605 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58233 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578118 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880794 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17028 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134453 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P153674 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P153671 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P153671 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P153674 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P153674 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P153671 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P18174 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 83769 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10939720 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10939724 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10939723 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10939721 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10939718 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10939722 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10802937 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10802936 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10939716 | - | - |
Rat Genome 230 2.0 Array | 1381474_PM_at | - | - |
Rat Genome 230 2.0 Array | 1381474_at | - | - |
Affymetrix HT Rat Focus Array | 1381474_at | - | - |
Affymetrix HT Rat Focus Array | 1381474_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0X00015566.rn.1 | TC0X00002477.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00015568.rn.1 | TC0X00002477.rn.1 | RTA TC |
Rat Expression Set 230 | 1381474_at | - | - |
Rat Expression Set 230 | 1381474_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10939716 | - | - |
Rat Genome U34 Set | rc_AI010322_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21038045 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363773 | - | - |
