Assay Details
Gene Symbol: Bcat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022400.1 | NP_071795.1 | 6 - 7 | 700 | 78 | |
XM_006229149.1 | XP_006229211.1 | 6 - 7 | 877 | 78 | ||
XM_008759413.1 | XP_008757635.1 | 6 - 7 | 1427 | 78 | ||
GenBank mRNA | BC061790.1 | - | 6 - 7 | 681 | 78 | |
U68417.1 | - | 6 - 7 | 700 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
