Assay Details
Gene Symbol: Ech1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022594.1 | NP_072116.1 | 8 - 9 | 739 | 63 | |
GenBank mRNA | BC060565.1 | - | 8 - 9 | 745 | 63 | |
BC062226.1 | - | 8 - 9 | 745 | 6890529 | 63 | |
FQ214121.1 | - | 8 - 9 | 741 | 63 | ||
FQ230435.1 | - | 8 - 9 | 752 | 63 | ||
U08976.1 | - | 8 - 9 | 739 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
