Assay Details
Gene Symbol: Prodh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038588.1 | NP_001033677.1 | 9 - 10 | 1206 | 94 | |
GenBank mRNA | BC158806.1 | - | 9 - 10 | 1206 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
