Assay Details
Gene Symbol: Prmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024363.1 | NP_077339.1 | 8 - 9 | 864 | 62 | |
XM_006229144.2 | XP_006229206.1 | 9 - 10 | 982 | 62 | ||
XM_006229145.2 | XP_006229207.1 | 8 - 9 | 1146 | 62 | ||
XM_006229146.2 | XP_006229208.1 | 9 - 10 | 1249 | 62 | ||
GenBank mRNA | BC078815.1 | - | 8 - 9 | 869 | 7106257 | 62 |
FQ219843.1 | - | 8 - 9 | 875 | 62 | ||
FQ220982.1 | - | 6 - 7 | 616 | 62 | ||
FQ228625.1 | - | 8 - 9 | 1145 | 62 | ||
U60882.1 | - | 8 - 9 | 864 | 62 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
