Assay Details
Gene Symbol: Prmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024363.1 | NP_077339.1 | 6 - 7 | 591 | 87 | |
XM_006229144.2 | XP_006229206.1 | 7 - 8 | 709 | 87 | ||
XM_006229145.2 | XP_006229207.1 | 6 - 7 | 873 | 87 | ||
XM_006229146.2 | XP_006229208.1 | 7 - 8 | 976 | 87 | ||
GenBank mRNA | BC078815.1 | - | 6 - 7 | 596 | 7106257 | 87 |
FQ213198.1 | - | 6 - 7 | 604 | 87 | ||
FQ219843.1 | - | 6 - 7 | 602 | 87 | ||
FQ220982.1 | - | 4 - 5 | 343 | 87 | ||
FQ228625.1 | - | 6 - 7 | 872 | 87 | ||
U60882.1 | - | 6 - 7 | 591 | 87 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
