Assay Details
Gene Symbol: Bckdha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012782.1 | NP_036914.1 | 2 - 3 | 292 | 81 | |
GenBank mRNA | BC161819.1 | - | 2 - 3 | 297 | 81 | |
CB579195.1 | - | 2 - 3 | 298 | 81 | ||
J02827.1 | - | 2 - 3 | 276 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
