Assay Details
Gene Symbol: Bckdha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012782.1 | NP_036914.1 | 4 - 5 | 490 | 72 | |
GenBank mRNA | BC089915.1 | - | 2 - 3 | 137 | 72 | |
BC161819.1 | - | 4 - 5 | 495 | 72 | ||
CB579195.1 | - | 4 - 5 | 496 | 72 | ||
J02827.1 | - | 4 - 5 | 474 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
