Assay Details
Gene Symbol: Ccne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100821.1 | NP_001094291.1 | 10 - 11 | 1119 | 75 | |
XM_006228904.2 | XP_006228966.1 | 10 - 11 | 1271 | 75 | ||
GenBank mRNA | BC162008.1 | - | 10 - 11 | 1146 | 75 | |
D14015.1 | - | 10 - 11 | 1136 | 75 | ||
DY561075.1 | - | 5 - 6 | 555 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
