Assay Details
Gene Symbol: Ccne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100821.1 | NP_001094291.1 | 2 - 3 | 191 | 110 | |
XM_006228904.2 | XP_006228966.1 | 2 - 3 | 343 | 224 | ||
GenBank mRNA | BC162008.1 | - | 2 - 3 | 218 | 224 | |
EV771022.1 | - | 2 - 3 | 225 | 110 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC162008.1, NM_001100821.1, XM_006228904.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
