Assay Details
Gene Symbol: Ccne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100821.1 | NP_001094291.1 | 3 - 4 | 277 | 74 | |
XM_006228904.2 | XP_006228966.1 | 3 - 4 | 429 | 74 | ||
GenBank mRNA | BC162008.1 | - | 3 - 4 | 304 | 74 | |
EV771022.1 | - | 3 - 4 | 311 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
