Assay Details
Gene Symbol: Kcnn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270701.1 | NP_001257630.1 | 7 - 8 | 1259 | 107 | |
NM_001270702.1 | NP_001257631.1 | 7 - 8 | 1172 | 107 | ||
NM_023021.2 | NP_075410.2 | 7 - 8 | 1259 | 107 | ||
XM_006228466.2 | XP_006228528.1 | 7 - 8 | 3019 | 107 | ||
XM_006228467.2 | XP_006228529.1 | 7 - 8 | 3020 | 107 | ||
XM_006228469.1 | XP_006228531.1 | 8 - 9 | 1241 | 107 | ||
XM_006228470.2 | XP_006228532.1 | 7 - 8 | 1089 | 107 | ||
XM_008758975.1 | XP_008757197.1 | 8 - 9 | 2914 | 107 | ||
XM_008758976.1 | XP_008757198.1 | 7 - 8 | 1438 | 107 | ||
XM_008758977.1 | XP_008757199.1 | 6 - 7 | 3420 | 107 | ||
XM_008758978.1 | XP_008757200.1 | 7 - 8 | 1078 | 107 | ||
GenBank mRNA | AB292802.1 | - | 5 - 6 | 734 | 107 | |
AF149250.1 | - | 7 - 8 | 1117 | 107 | ||
AF156554.1 | - | 7 - 8 | 1259 | 107 | ||
AF190458.1 | - | 7 - 8 | 1229 | 107 | ||
AJ133438.2 | - | 7 - 8 | 1117 | 107 | ||
BC128736.1 | - | 7 - 8 | 1210 | 8372655 | 107 | |
EU872449.1 | - | 7 - 8 | 1232 | 107 | ||
EU872450.1 | - | 7 - 8 | 1232 | 107 | ||
EU872451.1 | - | 7 - 8 | 1145 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
