Assay Details
Gene Symbol: Fxyd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031648.2 | NP_113836.2 | 2 - 3 | 146 | 76 | |
XM_006228838.2 | XP_006228900.1 | 3 - 4 | 366 | 76 | ||
XM_006228839.2 | XP_006228901.1 | 2 - 3 | 141 | 76 | ||
XM_008759184.1 | XP_008757406.1 | 2 - 3 | 155 | 76 | ||
GenBank mRNA | CB712570.1 | - | 2 - 3 | 146 | 76 | |
FM109156.1 | - | 2 - 3 | 100 | 76 | ||
FQ211349.1 | - | 2 - 3 | 157 | 76 | ||
FQ224131.1 | - | 2 - 3 | 109 | 76 | ||
FQ224338.1 | - | 2 - 3 | 192 | 76 | ||
U72246.1 | - | 1 - 2 | 60 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
