Assay Details
Gene Symbol: Aplp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100802.1 | NP_001094272.1 | 10 - 11 | 1400 | 71 | |
XM_008759179.1 | XP_008757401.1 | 9 - 10 | 1248 | 71 | ||
GenBank mRNA | BC161904.1 | - | 10 - 11 | 1400 | 71 | |
FQ213451.1 | - | 10 - 11 | 1412 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
