Assay Details
Gene Symbol: Aplp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100802.1 | NP_001094272.1 | 14 - 15 | 1680 | 103 | |
XM_008759179.1 | XP_008757401.1 | 13 - 14 | 1561 | 103 | ||
GenBank mRNA | BC161904.1 | - | 14 - 15 | 1680 | 103 | |
FQ212221.1 | - | 14 - 15 | 1670 | 103 | ||
FQ213451.1 | - | 14 - 15 | 1722 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
