Assay Details
Gene Symbol: Aplp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100802.1 | NP_001094272.1 | 1 - 2 | 196 | 107 | |
GenBank mRNA | BC161904.1 | - | 1 - 2 | 196 | 107 | |
FQ212221.1 | - | 1 - 2 | 156 | 107 | ||
FQ213451.1 | - | 1 - 2 | 208 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
