Assay Details
Gene Symbol: Sepw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013027.3 | NP_037159.4 | 3 - 4 | 197 | 111 | |
GenBank mRNA | BC087625.1 | - | 3 - 4 | 173 | 111 | |
BI279050.1 | - | 111 | ||||
DN931873.1 | - | 3 - 4 | 219 | 111 | ||
FQ212381.1 | - | 3 - 4 | 183 | 111 | ||
FQ212407.1 | - | 3 - 4 | 177 | 111 | ||
FQ215130.1 | - | 3 - 4 | 176 | 111 | ||
FQ215391.1 | - | 3 - 4 | 178 | 111 | ||
FQ216964.1 | - | 3 - 4 | 173 | 111 | ||
FQ217212.1 | - | 3 - 4 | 178 | 111 | ||
FQ217426.1 | - | 3 - 4 | 111 | |||
FQ217557.1 | - | 3 - 4 | 179 | 111 | ||
FQ217567.1 | - | 3 - 4 | 179 | 111 | ||
FQ217929.1 | - | 3 - 4 | 179 | 111 | ||
FQ221681.1 | - | 3 - 4 | 179 | 111 | ||
FQ223747.1 | - | 3 - 4 | 179 | 111 | ||
FQ224556.1 | - | 3 - 4 | 183 | 111 | ||
FQ224606.1 | - | 3 - 4 | 176 | 111 | ||
FQ224775.1 | - | 3 - 4 | 179 | 111 | ||
U25264.1 | - | 3 - 4 | 164 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
