Assay Details
Gene Symbol: Slc7a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053929.1 | NP_446381.1 | 1 - 2 | 36 | 67 | |
GenBank mRNA | AB029559.1 | - | 1 - 2 | 36 | 67 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AB029559.1, NM_053929.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
