Assay Details
Gene Symbol: Alad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012899.2 | NP_037031.1 | 1 - 2 | 87 | 117 | |
GenBank mRNA | BC061806.1 | - | 1 - 2 | 87 | 117 | |
FQ213585.1 | - | 1 - 2 | 229 | 117 | ||
FQ232546.1 | - | 1 - 2 | 63 | 117 | ||
FQ234245.1 | - | 1 - 2 | 63 | 117 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC061806.1, NM_012899.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
