Assay Details
Gene Symbol: Alad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012899.2 | NP_037031.1 | 5 - 6 | 566 | 69 | |
XM_006238234.2 | XP_006238296.1 | 5 - 6 | 1159 | 69 | ||
XM_006238235.1 | XP_006238297.1 | 5 - 6 | 656 | 69 | ||
GenBank mRNA | BC061806.1 | - | 5 - 6 | 566 | 69 | |
FQ210250.1 | - | 5 - 6 | 511 | 69 | ||
FQ213585.1 | - | 5 - 6 | 708 | 69 | ||
FQ218546.1 | - | 5 - 6 | 511 | 69 | ||
FQ219571.1 | - | 5 - 6 | 511 | 69 | ||
FQ231154.1 | - | 5 - 6 | 542 | 69 | ||
FQ232546.1 | - | 5 - 6 | 542 | 69 | ||
FQ233724.1 | - | 5 - 6 | 592 | 69 | ||
FQ234245.1 | - | 5 - 6 | 542 | 69 | ||
FQ234573.1 | - | 5 - 6 | 590 | 69 | ||
X04959.1 | - | 4 - 5 | 448 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
