Assay Details
Gene Symbol: Psip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175765.2 | NP_786941.1 | 2 - 3 | 247 | 118 | |
XM_006238353.2 | XP_006238415.1 | 3 - 4 | 471 | 118 | ||
XM_006238354.2 | XP_006238416.1 | 3 - 4 | 470 | 118 | ||
XM_008763779.1 | XP_008762001.1 | 3 - 4 | 470 | 118 | ||
GenBank mRNA | AB285525.1 | - | 150 | 118 | ||
AF339084.1 | - | 2 - 3 | 247 | 118 | ||
AF372090.1 | - | 2 - 3 | 247 | 118 | ||
BC093606.1 | - | 3 - 4 | 405 | 7317952 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
