Assay Details
Gene Symbol: Psip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175765.2 | NP_786941.1 | 3 - 4 | 384 | 131 | |
XM_006238353.2 | XP_006238415.1 | 4 - 5 | 608 | 131 | ||
XM_006238354.2 | XP_006238416.1 | 4 - 5 | 607 | 131 | ||
XM_008763779.1 | XP_008762001.1 | 4 - 5 | 607 | 131 | ||
GenBank mRNA | AB285525.1 | - | 287 | 131 | ||
AF339084.1 | - | 3 - 4 | 384 | 131 | ||
AF372090.1 | - | 3 - 4 | 384 | 131 | ||
BC093606.1 | - | 4 - 5 | 542 | 7317952 | 131 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
