Assay Details
Gene Symbol: Psip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175765.2 | NP_786941.1 | 7 - 8 | 721 | 159 | |
XM_006238353.2 | XP_006238415.1 | 8 - 9 | 945 | 159 | ||
XM_006238354.2 | XP_006238416.1 | 8 - 9 | 944 | 159 | ||
XM_008763779.1 | XP_008762001.1 | 8 - 9 | 944 | 159 | ||
GenBank mRNA | AB285525.1 | - | 624 | 159 | ||
AF339084.1 | - | 7 - 8 | 721 | 159 | ||
AF372090.1 | - | 7 - 8 | 721 | 159 | ||
BC093606.1 | - | 8 - 9 | 879 | 7317952 | 159 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
