Assay Details
Gene Symbol: Adamts5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198761.1 | NP_942056.1 | 3 - 4 | 1396 | 71 | |
GenBank mRNA | AY382879.1 | - | 3 - 4 | 1396 | 71 | |
FQ228283.1 | - | 1789 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
