Assay Details
Gene Symbol: Dcaf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173558.1 | NP_001167029.1 | 7 - 8 | 857 | 126 | |
XM_006241605.2 | XP_006241667.1 | 7 - 8 | 837 | 126 | ||
GenBank mRNA | BC158730.1 | - | 5 - 6 | 451 | 126 | |
FQ229670.1 | - | 5 - 6 | 516 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
