Assay Details
Gene Symbol: Trmu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135876.1 | NP_001129348.1 | 10 - 11 | 1172 | 70 | |
XM_006242164.2 | XP_006242226.1 | 9 - 10 | 1257 | 70 | ||
XM_008765766.1 | XP_008763988.1 | 9 - 10 | 882 | 70 | ||
GenBank mRNA | BC161991.1 | - | 9 - 10 | 1216 | 9032691 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
